Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTAGGTTCCTTAGCTAATCTAGGG[A/T]CCTTCATTTCATTATTAAGACCAGC
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
4930558K02Rik PubMed Links |
4930558K02Rik - RIKEN cDNA 4930558K02 gene | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001204904.1 | 1218 | Intron | NP_001191833.1 | |||
NM_001204905.1 | 1218 | Intron | NP_001191834.1 |
Pigc - phosphatidylinositol glycan anchor biosynthesis, class C | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001039045.1 | 1218 | UTR 3 | NP_001034134.1 | |||
NM_026078.2 | 1218 | UTR 3 | NP_080354.1 |