Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGCTTTCTTATTGATAGCTGATCT[A/C]ATTATGCTTGGTCTTGCTACACATG
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Xrn2 PubMed Links |
Xrn2 - 5'-3' exoribonuclease 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_011917.2 | 770 | Silent Mutation | NP_036047.2 |