Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGTAGACCTGTACACACACTCTGA[A/G]TACTACAACCACGCCATCGACTGGC
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Tshr PubMed Links |
Tshr - thyroid stimulating hormone receptor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001113404.1 | 1532 | Intron | NP_001106875.1 | |||
NM_011648.5 | 1532 | Silent Mutation | NP_035778.3 |