Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGAAGAGCTTCAACTCCAGGAAGGA[C/T]GGGCACTGGACAGCACAATATTTGG
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Cacnb4 PubMed Links |
Cacnb4 - calcium channel, voltage-dependent, beta 4 subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001037099.2 | Intron | NP_001032176.1 | ||||
NM_001285426.1 | Intron | NP_001272355.1 | ||||
NM_001285427.1 | Intron | NP_001272356.1 | ||||
NM_001285428.1 | Intron | NP_001272357.1 | ||||
NM_146123.3 | Intron | NP_666235.1 |