Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCTGACCAAAGTTTTCCAGATGCA[A/G]TTTCAGGCTGGGGCAAACTGTCCAT
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Grid2ip PubMed Links |
Grid2ip - glutamate receptor, ionotropic, delta 2 (Grid2) interacting protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001159321.1 | Intron | NP_001152793.1 | ||||
NM_133355.1 | Intron | NP_579933.1 |