Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGACTCAAAACACACAAACCATCTC[C/T]CCACAGTCCAGAGTTGTTTTACTCA
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Ntrk2 PubMed Links |
Ntrk2 - neurotrophic tyrosine kinase, receptor, type 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001025074.2 | Intron | NP_001020245.1 | ||||
NM_001282961.1 | Intron | NP_001269890.1 | ||||
NM_008745.3 | Intron | NP_032771.1 |