Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGATGTTGGTAGAGGTACCACTGGG[C/T]CCTGCAGAGGACTGGGCTTGCCTAG
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Tnr PubMed Links |
Tnr - tenascin R | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_022312.3 | Intron | NP_071707.2 |