Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTAAGTAGTTGTGTTGCTTGAAGG[C/T]GCAGGCCTCACCAAGTACCAAGCAT
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Cpped1 PubMed Links |
Cpped1 - calcineurin-like phosphoesterase domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_146067.3 | Intron | NP_666179.2 |