Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGAGAGCAAATGAGAGAAGATGTAC[G/A]GTTAGACTACTGCTCTGAGAAACAA
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Rhobtb3 PubMed Links |
Rhobtb3 - Rho-related BTB domain containing 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_028493.2 | Intron | NP_082769.1 |