Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATTAGTTGGCTCTTCGCCTCTTGG[A/C]TGATCTGTACCACTGCCAAGAATGA
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Gli3 PubMed Links |
Gli3 - GLI-Kruppel family member GLI3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_008130.2 | Intron | NP_032156.2 |