Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Chrd PubMed Links |
Chrd - chordin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278041.1 | 1764 | Silent Mutation | NP_001264970.1 | |||
NM_009893.2 | 1764 | Silent Mutation | NP_034023.1 |
Thpo - thrombopoietin | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |