Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGTGGTGCGAGGATTTCATGAGAC[A/C]AGGTATCCTATCAGCAGAGGGAATG
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Smyd3 PubMed Links |
Smyd3 - SET and MYND domain containing 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_027188.3 | Intron | NP_081464.1 |