Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTTCCCCGTTGGCCATGCACCTAG[C/T]GTGTGTCAGTTCCTGGAGCTGTGTC
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Fntb PubMed Links |
Fntb - farnesyltransferase, CAAX box, beta | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_145927.2 | 416 | Silent Mutation | NP_666039.1 |