Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAGATTCGAGTACAATGCTCGGTCT[A/C]TGGGGAGAGTTTTAAAGCTAAAGCT
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Rsrc1 PubMed Links |
Rsrc1 - arginine/serine-rich coiled-coil 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_025822.3 | Intron | NP_080098.1 |