Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCAAATGTCCATAGTATATCCAATA[C/T]GAAGCAATTTGAGTGAACAGTTTTT
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Ccnb3 PubMed Links |
Ccnb3 - cyclin B3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_183015.3 | Intron | NP_898836.2 |