Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCGGGATCTGGGCGAGCTCTGAGCA[C/T]TCATCCCAACACGCTCGCTCTTTAC
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Osr2 PubMed Links |
Osr2 - odd-skipped related 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_054049.2 | 134 | UTR 5 | NP_473390.1 |