Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAACAGGGTTGATTAGAATGTATAG[A/G]AGGTCCTCCTCCCTAAAGCTGGAGG
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Ptn PubMed Links |
Ptn - pleiotrophin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_008973.2 | Intron | NP_032999.1 |