Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCTGGGCCTGGCTGCCCACCCCTA[C/T]GATCTCAGCTACTCAAATTCTCTTA
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Cltc PubMed Links |
Cltc - clathrin, heavy polypeptide (Hc) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001003908.1 | Intron | NP_001003908.1 |