Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCTTATAATTGTGACTTTATTTAC[A/T]TGTAACTTTGCTGTCTTTTCTTTAA
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Dennd1b PubMed Links |
Dennd1b - DENN/MADD domain containing 1B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001166501.1 | Intron | NP_001159973.1 | ||||
NM_181347.3 | Intron | NP_851992.1 |