Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACATAGCAGAAACCTCCTTTCTAT[A/G]AGCACTGCAGCCATCAGAACCCAAG
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Ralgapa2 PubMed Links |
Ralgapa2 - Ral GTPase activating protein, alpha subunit 2 (catalytic) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001033348.3 | Intron | NP_001028520.2 |