Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATTAGAATACACAGGGGTTATCTT[C/T]CAGAGTATCACAGAATGTAATTGTT
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Mmd PubMed Links |
Mmd - monocyte to macrophage differentiation-associated | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_026178.2 | Intron | NP_080454.1 |