Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Arhgap36 PubMed Links |
Arhgap36 - Rho GTPase activating protein 36 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001081123.1 | Intron | NP_001074592.1 |
Gm14696 - predicted gene 14696 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |