Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGTGTTTCCGGAGACACAGGGAATG[A/C]TGGGGTCAGACACATAGATGACTCC
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Tns3 PubMed Links |
Tns3 - tensin 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001083587.1 | 2836 | Missense Mutation | NP_001077056.1 |