Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGGCATCTGACACTGGAAACCACTG[A/G]TCTGCTAAGCTGGCTGGAGTGTAAG
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Skap1 PubMed Links |
Skap1 - src family associated phosphoprotein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001033186.3 | Intron | NP_001028358.1 | ||||
NM_001177898.1 | Intron | NP_001171369.1 | ||||
NM_001177899.1 | Intron | NP_001171370.1 |