Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Cxxc4 PubMed Links |
Cxxc4 - CXXC finger 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001004367.4 | Intron | NP_001004367.2 |
Mir1895 - microRNA 1895 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |