Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGGAGAAAGGGTTAACAGAAGGAA[A/T]GAGAAAATGTTACACATTTTCCCTC
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Qrfpr PubMed Links |
Qrfpr - pyroglutamylated RFamide peptide receptor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_198192.2 | Intron | NP_937835.1 |