Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTCCAGATCAGCTGCGCTGGCCAG[C/T]CCATGATGAGCCCCTCCAGGCAAAC
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Nckap5 PubMed Links |
Nckap5 - NCK-associated protein 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001081756.1 | Intron | NP_001075225.1 | ||||
NM_172484.3 | Intron | NP_766072.2 | ||||
NM_176957.3 | Intron | NP_795931.2 |