Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Chrna6 PubMed Links |
Chrna6 - cholinergic receptor, nicotinic, alpha polypeptide 6 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Chrnb3 - cholinergic receptor, nicotinic, beta polypeptide 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_027454.4 | 1236 | Silent Mutation | NP_081730.1 | |||
NM_173212.4 | 1236 | Silent Mutation | NP_775304.1 |