Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCACGGCAAACCGGACGACACTTGG[A/G]GGCGGCTCAGGCGAAGGAGTGGGCA
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Ptch1 PubMed Links |
Ptch1 - patched homolog 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_008957.2 | 3561 | Silent Mutation | NP_032983.1 |