Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGTCAACATCCGCTAAAGAAGCGT[A/G]GGACACTCTGCATATTGTCTAGTTC
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Aldh1l2 PubMed Links |
Aldh1l2 - aldehyde dehydrogenase 1 family, member L2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_153543.2 | 1552 | Missense Mutation | NP_705771.2 |