Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCTTATTCCCTAGTGATCTCATCA[A/G]TTTGCTAAGGAAGACTGTCTGCAAA
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Hiatl1 PubMed Links |
Hiatl1 - hippocampus abundant transcript-like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001083901.1 | 1208 | Silent Mutation | NP_001077370.1 | |||
NM_133680.3 | 1208 | Silent Mutation | NP_598441.3 |