Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTCCAGCACATGCCTGCTGTCCAC[T/C]ATTTCCATCATTGAGTCACAAGACA
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Dtd1 PubMed Links |
Dtd1 - D-tyrosyl-tRNA deacylase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_025314.3 | 1127 | UTR 3 | NP_079590.1 |