Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTCCCAAAGGACCAAAAGGAGACCC[A/G]GGAGAGCCTGGACCAGCTGGGCCCA
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Col23a1 PubMed Links |
Col23a1 - collagen, type XXIII, alpha 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_153393.2 | 1269 | Silent Mutation | NP_700442.2 |