Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTGTGAACTGCAAGGCAGCTTTCA[G/T]AAAGCATCGCCCACCCCACCCCCTG
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Adamts2 PubMed Links |
Adamts2 - a disintegrin-like and metallopeptidase (reprolysin type) with thrombospondin type 1 motif, 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001277305.1 | Intron | NP_001264234.1 | ||||
NM_175643.3 | Intron | NP_783574.1 |