Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTTCATAGGATCTAGGAAAAGGGC[A/G]TGTAGAATGTTTCCAGAACCTTCGA
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Npas2 PubMed Links |
Npas2 - neuronal PAS domain protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_008719.2 | Intron | NP_032745.2 |