Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGTAATATGCATGGAAGTGTTTTCG[A/C]ACAATTTCCACACACTTTCCTCCTC
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Khdrbs2 PubMed Links |
Khdrbs2 - KH domain containing, RNA binding, signal transduction associated 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_133235.2 | Intron | NP_573498.1 |