Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGAAGGGCAGACTACAAAGCACCC[A/G]GGGGTAAAAGGGCAGCAAGGAAGGG
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Nisch PubMed Links |
Nisch - nischarin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_022656.2 | Intron | NP_073147.2 |