Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACCATGTTTAACCTCAGTGGCTTA[G/T]GAGCTTCCAGATATTCCCTTTTCTC
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Dtnb PubMed Links |
Dtnb - dystrobrevin, beta | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001162465.1 | Intron | NP_001155937.1 | ||||
NM_007886.2 | Intron | NP_031912.2 |