Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGCTGTGTGTACATGCTTTAAAAC[A/G]TATCTTTCACTCGAAACCATATGCT
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Trpm3 PubMed Links |
Trpm3 - transient receptor potential cation channel, subfamily M, member 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001035239.2 | Intron | NP_001030316.1 | ||||
NM_001035240.2 | Intron | NP_001030317.1 | ||||
NM_001035241.2 | Intron | NP_001030318.1 | ||||
NM_001035242.1 | Intron | NP_001030319.1 | ||||
NM_001035243.2 | Intron | NP_001030320.1 | ||||
NM_001035244.1 | Intron | NP_001030321.1 | ||||
NM_001035245.1 | Intron | NP_001030322.1 | ||||
NM_001035246.1 | Intron | NP_001030323.1 | ||||
NM_177341.4 | Intron | NP_796315.2 |