Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCCCCCTCTCTGCTCGCCATGACC[A/G]GCATCTTTGTTACTATCACTGGTTA
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Sdk1 PubMed Links |
Sdk1 - sidekick homolog 1 (chicken) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_177879.5 | Intron | NP_808547.3 |