Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCATTCTCCTAAGTTAAAATAGTCA[A/G]ACCTATTGCATAGCTTCTCCCACTT
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Trhde PubMed Links |
Trhde - TRH-degrading enzyme | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_146241.2 | Intron | NP_666353.1 |