Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAAATCTGTGAGGAGTGGTTGTGGA[G/C]ACCAGTATCTTTTAGAGGATTGTCT
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Lrba PubMed Links |
Lrba - LPS-responsive beige-like anchor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001077687.1 | Intron | NP_001071155.1 | ||||
NM_001077688.1 | Intron | NP_001071156.1 | ||||
NM_030695.2 | Intron | NP_109620.2 |