Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Bcap29 PubMed Links |
Bcap29 - B cell receptor associated protein 29 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Dus4l - dihydrouridine synthase 4-like (S. cerevisiae) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_028002.2 | 1107 | Missense Mutation | NP_082278.1 |