Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Gm10258 PubMed Links |
Gm10258 - predicted gene 10258 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Mecom - MDS1 and EVI1 complex locus | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_007963.2 | Intron | NP_031989.2 | ||||
NM_021442.2 | Intron | NP_067417.1 |