Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Gm26559 PubMed Links |
Gm26559 - predicted gene, 26559 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Map1b - microtubule-associated protein 1B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_008634.2 | 328 | Silent Mutation | NP_032660.2 |