Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGTATCTGCGGTCAGCAATACCAC[A/C]GCAAAAGCAGCCTCCTTGCTCTTCA
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Arhgef28 PubMed Links |
Arhgef28 - Rho guanine nucleotide exchange factor (GEF) 28 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_012026.2 | Intron | NP_036156.2 |