Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATTGGTATCAAGAATGTCTGGGAA[A/G]AGATGGGATAATTTTTTCTCATGAT
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Arsb PubMed Links |
Arsb - arylsulfatase B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_009712.3 | Intron | NP_033842.3 |