Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCACCTTCATCTCCAAGTTCAAAAT[T/C]AAAGTGCAGAAGAAGCCATTTACCA
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
1500026H17Rik PubMed Links |
1500026H17Rik - RIKEN cDNA 1500026H17 gene | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Scyl2 - SCY1-like 2 (S. cerevisiae) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_198021.2 | Intron | NP_932138.1 |