Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCGCAATTCCCGGCCTTGGTGGCA[C/G]TTGTTTAGTGAGCGGACGAGGTACT
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Serpina9 PubMed Links |
Serpina9 - serine (or cysteine) peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), member 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_027997.2 | 441 | Missense Mutation | NP_082273.1 |