Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCACTCTGCTGGGAATAGATCAAAC[C/G]AATCCTGTCCCGGGGAAAGGCCTAA
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Galnt14 PubMed Links |
Galnt14 - UDP-N-acetyl-alpha-D-galactosamine:polypeptide N-acetylgalactosaminyltransferase 14 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_027864.2 | Intron | NP_082140.2 |